| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10009716.hg.1 |
ENST00000265277.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10009716.hg.1 |
ENST00000369452.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10009716.hg.1 |
ENST00000489390.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10009716.hg.1 |
NM_007373.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10009716.hg.1 |
TC10000809.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000265277.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000265277.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000265277.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000265277.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000265277.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000265277.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000265277.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000265277.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000265277.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000265277.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000265277.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000265277.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000265277.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000265277.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000265277.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000369452.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000369452.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000369452.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000369452.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000369452.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000369452.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000369452.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000369452.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000369452.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000369452.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000369452.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000369452.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000369452.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000369452.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000369452.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000369452.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000369452.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000369452.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000451838.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000451838.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000451838.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000451838.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000451838.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000451838.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000451838.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000451838.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000451838.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000451838.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000480155.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000480155.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000480155.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000480155.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000480155.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000489390.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000489390.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000489390.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000489390.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000489390.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000489390.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000489390.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000489783.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000489783.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000489783.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000489783.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000489783.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000497305.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000497305.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
ENST00000497305.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
NM_001269039.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
NM_001269039.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
NM_001269039.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
NM_001269039.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
NM_001269039.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
NM_001269039.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
NM_001269039.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
NM_001269039.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
NM_001269039.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
NM_001269039.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
NM_001269039.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
NM_001269039.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
NM_001269039.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
NM_001269039.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
NM_001269039.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
NM_007373.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
NM_007373.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
NM_007373.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
NM_007373.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
NM_007373.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
NM_007373.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
NM_007373.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
NM_007373.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
NM_007373.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
NM_007373.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
NM_007373.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
NM_007373.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
NM_007373.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
NM_007373.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
NM_007373.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
NM_007373.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
NM_007373.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008908.hg.1 |
NM_007373.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000160936.hg.1 |
ENST00000265277.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000160936.hg.1 |
ENST00000369452.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000160936.hg.1 |
ENST00000451838.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000160936.hg.1 |
ENST00000489390.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000160936.hg.1 |
NM_001269039.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000160936.hg.1 |
NM_007373.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000160936.hg.1 |
ENST00000489390.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000160936.hg.1 |
ENST00000265277.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000160936.hg.1 |
ENST00000369452.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000160936.hg.1 |
ENST00000451838.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000160936.hg.1 |
NM_001269039.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000160936.hg.1 |
TC1000008908.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000160936.hg.1 |
NM_007373.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000265277.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000265277.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000265277.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000265277.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000265277.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000265277.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000265277.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000265277.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000265277.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000265277.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000265277.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000265277.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000265277.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000265277.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000265277.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000369452.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000369452.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000369452.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000369452.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000369452.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000369452.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000369452.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000369452.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000369452.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000369452.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000369452.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000369452.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000369452.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000369452.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000369452.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000369452.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000369452.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000369452.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000451838.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000451838.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000451838.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000451838.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000451838.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000451838.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000451838.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000451838.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000451838.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000451838.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000480155.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000480155.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000480155.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000480155.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000480155.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000489390.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000489390.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000489390.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000489390.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000489390.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000489390.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000489390.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000489783.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000489783.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000489783.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000489783.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000489783.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000497305.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000497305.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
ENST00000497305.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
NM_001269039.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
NM_001269039.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
NM_001269039.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
NM_001269039.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
NM_001269039.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
NM_001269039.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
NM_001269039.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
NM_001269039.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
NM_001269039.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
NM_001269039.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
NM_001269039.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
NM_001269039.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
NM_001269039.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
NM_001269039.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
NM_001269039.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
NM_007373.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
NM_007373.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
NM_007373.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
NM_007373.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
NM_007373.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
NM_007373.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
NM_007373.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
NM_007373.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
NM_007373.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
NM_007373.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
NM_007373.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
NM_007373.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
NM_007373.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
NM_007373.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
NM_007373.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
NM_007373.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
NM_007373.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008908.hg.1 |
NM_007373.3 |
Refseq |