Assay Details
| Gene Symbol: PRSS27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK126510.1 | - | 1 | 2967 | 61 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000302641.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000302641.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000302641.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000302641.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000302641.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000302641.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000302641.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000302641.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000543965.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000543965.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000543965.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000543965.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000543965.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000543965.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000562249.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000562249.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000562249.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000562249.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000562249.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000562249.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000562249.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000565903.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000565903.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000565903.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000565903.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000565903.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000565903.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000565903.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000565903.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000566492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000566492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000566492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000566492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000566492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000566492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | NM_031948.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | NM_031948.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | NM_031948.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | NM_031948.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | NM_031948.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | NM_031948.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | NM_031948.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | NM_031948.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | NM_031948.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000302641.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000302641.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000302641.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000302641.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000302641.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000302641.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000302641.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000302641.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000543965.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000543965.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000543965.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000543965.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000543965.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000543965.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000562249.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000562249.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000562249.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000562249.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000562249.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000562249.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000562249.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000565903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000565903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000565903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000565903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000565903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000565903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000565903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000565903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000566492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000566492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000566492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000566492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000566492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000566492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | NM_031948.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | NM_031948.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | NM_031948.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | NM_031948.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | NM_031948.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | NM_031948.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | NM_031948.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | NM_031948.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | NM_031948.4 | Refseq |

PRSS27
Genomic Map