| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013035.hg.1 |
ENST00000413292.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013035.hg.1 |
ENST00000413292.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013035.hg.1 |
ENST00000413292.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013035.hg.1 |
ENST00000413292.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000528530.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000528530.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000528530.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000528530.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000532850.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000532850.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000532850.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000532850.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
ENST00000532850.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11007507.hg.1 |
ENST00000164133.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11007507.hg.1 |
ENST00000359279.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11007507.hg.1 |
ENST00000527441.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11007507.hg.1 |
NM_006244.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11007507.hg.1 |
TC11000611.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000164133.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000528530.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000528530.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000528530.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000528530.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000532850.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000532850.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000532850.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000532850.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
ENST00000532850.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013036.hg.1 |
NM_006244.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100201134.hg.1 |
ENST00000164133.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100201134.hg.1 |
ENST00000532850.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100201134.hg.1 |
NM_006244.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100201134.hg.1 |
ENST00000164133.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100201134.hg.1 |
ENST00000532850.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100201134.hg.1 |
NM_006244.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100201134.hg.1 |
TC1100013036.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100201131.hg.1 |
TC1100013036.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100201135.hg.1 |
ENST00000164133.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100201135.hg.1 |
ENST00000532850.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100201135.hg.1 |
NM_006244.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100201135.hg.1 |
ENST00000164133.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100201135.hg.1 |
ENST00000532850.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100201135.hg.1 |
NM_006244.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100201135.hg.1 |
TC1100013036.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1100013035.hg.1 |
ENST00000413292.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013035.hg.1 |
ENST00000413292.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013035.hg.1 |
ENST00000413292.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013035.hg.1 |
ENST00000413292.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100201133.hg.1 |
ENST00000164133.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100201133.hg.1 |
NM_006244.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100201133.hg.1 |
ENST00000164133.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100201133.hg.1 |
NM_006244.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100201133.hg.1 |
TC1100013036.hg.1 |
Clariom D TC |