Assay Details
| Gene Symbol: EPB41L4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018424.3 | NP_060894.2 | 7 - 8 | 1267 | 116 | |
| NM_019114.4 | NP_061987.3 | 7 - 8 | 1267 | 116 | ||
| XM_011518790.1 | XP_011517092.1 | 7 - 8 | 764 | 116 | ||
| XM_011518791.2 | XP_011517093.1 | 7 - 8 | 749 | 116 | ||
| XM_011518792.2 | XP_011517094.1 | 7 - 8 | 749 | 116 | ||
| XM_011518794.2 | XP_011517096.1 | 7 - 8 | 749 | 116 | ||
| XM_017014813.1 | XP_016870302.1 | 7 - 8 | 749 | 116 | ||
| XM_017014814.1 | XP_016870303.1 | 7 - 8 | 749 | 116 | ||
| XM_017014815.1 | XP_016870304.1 | 7 - 8 | 749 | 116 | ||
| GenBank mRNA | AB032179.3 | - | 7 - 8 | 1267 | 116 | |
| AF153416.1 | - | 7 - 8 | 1039 | 116 | ||
| AF153418.1 | - | 7 - 8 | 1039 | 116 | ||
| AK000388.1 | - | 7 - 8 | 651 | 116 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001466.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of gene expression
regulation of keratinocyte proliferation
actomyosin structure organization
wound healing
positive regulation of cell adhesion
positive regulation of keratinocyte migration
structural constituent of cytoskeleton
cytoskeletal protein binding
positive regulation of gene expression
regulation of keratinocyte proliferation
actomyosin structure organization
wound healing
positive regulation of cell adhesion
positive regulation of keratinocyte migration
structural constituent of cytoskeleton
cytoskeletal protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | ENST00000374557.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | ENST00000374557.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | ENST00000374557.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | ENST00000374557.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | ENST00000374557.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | ENST00000374557.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | ENST00000374557.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | ENST00000374557.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | ENST00000374557.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | ENST00000374557.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | ENST00000374557.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | ENST00000374557.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | ENST00000374557.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | ENST00000374557.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | ENST00000374557.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | ENST00000374557.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | ENST00000374557.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | ENST00000374557.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | NM_018424.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | NM_018424.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | NM_018424.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | NM_018424.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | NM_018424.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | NM_018424.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | NM_018424.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | NM_018424.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | NM_018424.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | NM_018424.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | NM_018424.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | NM_018424.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | NM_018424.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | NM_018424.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | NM_018424.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | NM_018424.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | NM_018424.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | NM_018424.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018119.hg.1 | ENST00000262536.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018119.hg.1 | ENST00000311609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018119.hg.1 | ENST00000374557.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018119.hg.1 | ENST00000374566.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018119.hg.1 | NM_018424.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018119.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018119.hg.1 | TC09001466.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018120.hg.1 | ENST00000262536.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018120.hg.1 | ENST00000311609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018120.hg.1 | ENST00000374557.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018120.hg.1 | ENST00000374566.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018120.hg.1 | NM_018424.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018120.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018120.hg.1 | TC09001466.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0900175924.hg.1 | ENST00000374557.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900175924.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900175924.hg.1 | NM_018424.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900175924.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900175924.hg.1 | ENST00000374557.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900175924.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900175924.hg.1 | NM_018424.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900175924.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900175924.hg.1 | TC0900011160.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | ENST00000374557.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | ENST00000374557.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | ENST00000374557.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | ENST00000374557.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | ENST00000374557.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | ENST00000374557.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | ENST00000374557.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | ENST00000374557.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | ENST00000374557.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | ENST00000374557.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | ENST00000374557.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | ENST00000374557.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | ENST00000374557.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | ENST00000374557.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | ENST00000374557.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | ENST00000374557.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | ENST00000374557.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | ENST00000374557.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | ENST00000374566.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | NM_018424.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | NM_018424.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | NM_018424.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | NM_018424.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | NM_018424.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | NM_018424.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | NM_018424.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | NM_018424.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | NM_018424.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | NM_018424.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | NM_018424.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | NM_018424.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | NM_018424.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | NM_018424.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | NM_018424.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | NM_018424.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | NM_018424.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | NM_018424.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | NM_019114.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011160.hg.1 | NM_019114.4 | Refseq |

EPB41L4B
Genomic Map