Assay Details
| Gene Symbol: MAP3K6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004672.4 | NP_004663.3 | 2 - 3 | 748 | 99 | |
| XM_017002772.1 | XP_016858261.1 | 2 - 3 | 1919 | 99 | ||
| GenBank mRNA | AB167411.1 | - | 2 - 3 | 479 | 99 | |
| AI582755.1 | - | 2 - 3 | 225 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

MAP3K6
Genomic Map