Assay Details
| Gene Symbol: ZACN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_180990.3 | NP_851321.2 | 2 - 3 | 305 | 62 | |
| GenBank mRNA | AB223030.1 | - | 2 - 3 | 219 | 62 | |
| AF512521.1 | - | 2 - 3 | 259 | 62 | ||
| AK122638.1 | - | 2 - 3 | 305 | 62 | ||
| BC110597.1 | - | 2 - 3 | 259 | 40023955 | 62 | |
| DB119720.1 | - | 2 - 3 | 305 | 62 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000865.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
serotonin receptor signaling pathway
synaptic transmission, cholinergic
response to zinc ion
ion transmembrane transport
cation transmembrane transport
acetylcholine-activated cation-selective channel activity
G-protein coupled serotonin receptor activity
serotonin-activated cation-selective channel activity
zinc ion binding
ligand-gated ion channel activity
acetylcholine receptor activity
acetylcholine binding
serotonin receptor signaling pathway
synaptic transmission, cholinergic
response to zinc ion
ion transmembrane transport
cation transmembrane transport
acetylcholine-activated cation-selective channel activity
G-protein coupled serotonin receptor activity
serotonin-activated cation-selective channel activity
zinc ion binding
ligand-gated ion channel activity
acetylcholine receptor activity
acetylcholine binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17011287.hg.1 | ENST00000334586.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17011287.hg.1 | NM_180990.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17011287.hg.1 | TC17000865.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000334586.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000334586.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000334586.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000334586.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000334586.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000334586.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000334586.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000334586.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000334586.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000334586.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000334586.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000334586.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000421794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000421794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000421794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000421794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000421794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000421794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000421794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000421794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000421794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000425015.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000425015.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000425015.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000425015.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000425015.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000425015.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000425015.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000425015.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000425015.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000425015.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000425015.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000425015.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000425015.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000425015.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000425015.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000425015.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000524242.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000524242.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000524242.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000524242.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000524242.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000524242.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000524242.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000590045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000590045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000590045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000590045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000590045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000590045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000590045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000590045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000590045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000590045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000590045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000590045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000591500.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000591500.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000591500.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000591500.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | ENST00000591500.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | NM_180990.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | NM_180990.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | NM_180990.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | NM_180990.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | NM_180990.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | NM_180990.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | NM_180990.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | NM_180990.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | NM_180990.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | NM_180990.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | NM_180990.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | NM_180990.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008932.hg.1 | NM_180990.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700167271.hg.1 | ENST00000334586.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700167271.hg.1 | ENST00000425015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700167271.hg.1 | ENST00000524242.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700167271.hg.1 | ENST00000591500.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700167271.hg.1 | NM_180990.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700167271.hg.1 | TC1700008932.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700167271.hg.1 | ENST00000334586.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700167271.hg.1 | ENST00000425015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700167271.hg.1 | ENST00000524242.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700167271.hg.1 | ENST00000591500.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700167271.hg.1 | NM_180990.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000334586.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000334586.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000334586.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000334586.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000334586.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000334586.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000334586.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000334586.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000334586.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000334586.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000334586.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000334586.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000421794.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000421794.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000421794.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000421794.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000421794.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000421794.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000421794.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000421794.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000421794.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000425015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000425015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000425015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000425015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000425015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000425015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000425015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000425015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000425015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000425015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000425015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000425015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000425015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000425015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000425015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000425015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000524242.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000524242.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000524242.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000524242.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000524242.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000524242.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000524242.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000590045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000590045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000590045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000590045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000590045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000590045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000590045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000590045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000590045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000590045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000590045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000590045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000591500.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000591500.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000591500.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000591500.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | ENST00000591500.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | NM_180990.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | NM_180990.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | NM_180990.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | NM_180990.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | NM_180990.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | NM_180990.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | NM_180990.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | NM_180990.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | NM_180990.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | NM_180990.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | NM_180990.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | NM_180990.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008932.hg.1 | NM_180990.3 | Refseq |

ZACN
Genomic Map