Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ATR interacting protein | ATRIP | Sense | Chr. 3 - 48446710 - 48465655 on NCBI Build GRCh38 | ||
| Downstream | three prime repair exonuclease 1 | TREX1 |
AGS1, CRV, DRN3, HERNS |
93.0 | Sense | Chr. 3 - 48465520 - 48467645 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map