Assay Details
| Gene Symbol: CLPX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006660.4 | NP_006651.2 | 1 - 2 | 344 | 90 | |
| NR_133680.1 | 1 - 2 | 344 | 90 | |||
| XM_011521164.2 | XP_011519466.1 | 1 - 2 | 328 | 90 | ||
| GenBank mRNA | AJ006267.3 | - | 1 - 2 | 149 | 90 | |
| AK292316.1 | - | 1 - 2 | 270 | 90 | ||
| AK308133.1 | - | 1 - 2 | 264 | 90 | ||
| AK310614.1 | - | 1 - 2 | 266 | 90 | ||
| AL136922.1 | - | 1 - 2 | 247 | 90 | ||
| BC130373.1 | - | 1 - 2 | 140 | 40146404 | 90 | |
| BC136487.1 | - | 1 - 2 | 140 | 9020476 | 90 | |
| BG717994.1 | - | 1 - 2 | 349 | 90 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15001561.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR15014901.hg.1 | ENST00000300107.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014901.hg.1 | ENST00000546194.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014901.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014901.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014901.hg.1 | TC15001561.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000558103.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000558103.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000558103.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000558103.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000559218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000559218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000559218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000559218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000560166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000560166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000560166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014900.hg.1 | ENST00000300107.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014900.hg.1 | ENST00000546194.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014900.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014900.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014900.hg.1 | TC15001561.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014902.hg.1 | ENST00000300107.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014902.hg.1 | ENST00000546194.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014902.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014902.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014902.hg.1 | TC15001561.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1500172322.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172322.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172322.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172322.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500172322.hg.1 | TC1500009785.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500172322.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172322.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172322.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172322.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000558103.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000558103.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000558103.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000558103.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000559218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000559218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000559218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000559218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000560166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000560166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000560166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500172319.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172319.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172319.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172319.hg.1 | ENST00000559218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172319.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500172319.hg.1 | TC1500009785.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500172319.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172319.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172319.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172319.hg.1 | ENST00000559218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172319.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500172321.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172321.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172321.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172321.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500172321.hg.1 | TC1500009785.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500172321.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172321.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172321.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172321.hg.1 | NM_006660.4 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 202862 | - | - |

CLPX
Genomic Map