Assay Details
| Gene Symbol: CLPX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006660.4 | NP_006651.2 | 2 - 3 | 508 | 106 | |
| NR_133680.1 | 2 - 3 | 508 | 106 | |||
| XM_011521164.2 | XP_011519466.1 | 2 - 3 | 492 | 106 | ||
| GenBank mRNA | AJ006267.3 | - | 2 - 3 | 313 | 106 | |
| AK292316.1 | - | 2 - 3 | 434 | 106 | ||
| AK308133.1 | - | 2 - 3 | 428 | 106 | ||
| AK310614.1 | - | 2 - 3 | 430 | 106 | ||
| AL136922.1 | - | 2 - 3 | 411 | 106 | ||
| BC130373.1 | - | 2 - 3 | 304 | 40146404 | 106 | |
| BC136487.1 | - | 2 - 3 | 304 | 9020476 | 106 | |
| BG717994.1 | - | 2 - 3 | 514 | 107 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15001561.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000558103.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000558103.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000558103.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000558103.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000559218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000559218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000559218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000559218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000560166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000560166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | ENST00000560166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014899.hg.1 | ENST00000300107.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014899.hg.1 | ENST00000546194.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014899.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014899.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014899.hg.1 | TC15001561.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000558103.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000558103.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000558103.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000558103.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000559218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000559218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000559218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000559218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000560166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000560166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | ENST00000560166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009785.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500172317.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172317.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172317.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172317.hg.1 | ENST00000559218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172317.hg.1 | NM_006660.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500172317.hg.1 | TC1500009785.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500172317.hg.1 | ENST00000300107.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172317.hg.1 | ENST00000558958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172317.hg.1 | ENST00000559152.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172317.hg.1 | ENST00000559218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172317.hg.1 | NM_006660.4 | Refseq |

CLPX
Genomic Map