| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009982.hg.1 |
ENST00000559052.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01038983.hg.1 |
ENST00000373116.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01038983.hg.1 |
NM_031280.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01038983.hg.1 |
TC01002511.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01038985.hg.1 |
ENST00000373116.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01038985.hg.1 |
NM_031280.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01038985.hg.1 |
TC01002511.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01038987.hg.1 |
ENST00000373116.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01038987.hg.1 |
NM_031280.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01038987.hg.1 |
TC01002511.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000373116.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000373116.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000373116.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000373116.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000373116.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000373116.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000373116.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000373116.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000373116.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000373116.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000373116.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000462067.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000462067.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000462067.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000462067.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000462067.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000477040.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000477040.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000477040.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000488606.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000488606.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000488606.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000488606.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
ENST00000488606.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
NM_031280.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
NM_031280.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
NM_031280.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
NM_031280.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
NM_031280.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
NM_031280.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
NM_031280.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
NM_031280.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
NM_031280.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013723.hg.1 |
NM_031280.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01038984.hg.1 |
ENST00000373116.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01038984.hg.1 |
NM_031280.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01038984.hg.1 |
TC01002511.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1500009982.hg.1 |
ENST00000559052.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100202304.hg.1 |
ENST00000373116.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100202304.hg.1 |
NM_031280.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100202304.hg.1 |
ENST00000373116.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100202304.hg.1 |
NM_031280.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100202304.hg.1 |
TC0100013723.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100202303.hg.1 |
ENST00000373116.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100202303.hg.1 |
NM_031280.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100202303.hg.1 |
ENST00000373116.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100202303.hg.1 |
NM_031280.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100202303.hg.1 |
TC0100013723.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000373116.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000373116.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000373116.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000373116.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000373116.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000373116.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000373116.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000373116.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000373116.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000373116.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000373116.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000462067.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000462067.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000462067.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000462067.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000462067.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000477040.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000477040.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000477040.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000488606.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000488606.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000488606.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000488606.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
ENST00000488606.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
NM_031280.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
NM_031280.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
NM_031280.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
NM_031280.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
NM_031280.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
NM_031280.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
NM_031280.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
NM_031280.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
NM_031280.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013723.hg.1 |
NM_031280.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100202306.hg.1 |
ENST00000373116.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100202306.hg.1 |
ENST00000462067.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100202306.hg.1 |
NM_031280.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100202306.hg.1 |
ENST00000373116.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100202306.hg.1 |
ENST00000462067.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100202306.hg.1 |
NM_031280.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100202306.hg.1 |
TC0100013723.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100202308.hg.1 |
ENST00000373116.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100202308.hg.1 |
NM_031280.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100202308.hg.1 |
ENST00000373116.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100202308.hg.1 |
TC0100013723.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100202308.hg.1 |
NM_031280.3 |
Refseq |