Assay Details
| Gene Symbol: EMX2OS | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK123533.1 | - | 1 - 2 | 149 | 79 | ||
| AY117413.1 | - | LIT2056 EMX2OS |
2 - 3 | 405 | 79 | ||
| BC040186.2 | - | 1 - 2 | 243 | 79 | |||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000423419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000423419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000423419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000423419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000424371.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000424371.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000424371.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000424371.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000450314.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000450314.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000450314.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000450314.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000450314.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000450314.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000450314.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000450314.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000611404.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000611404.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000611404.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000423419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000423419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000423419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000423419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000424371.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000424371.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000424371.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000424371.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000450314.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000450314.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000450314.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000450314.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000450314.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000450314.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000450314.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000450314.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000611404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000611404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000611404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | NR_002791.2 | Refseq |

EMX2OS
Genomic Map