| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
ENST00000369073.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
ENST00000369073.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
ENST00000369073.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
ENST00000369073.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
ENST00000369073.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
ENST00000369073.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
ENST00000369073.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
ENST00000369073.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
ENST00000369073.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
ENST00000369073.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
ENST00000369073.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
ENST00000398250.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
ENST00000398250.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
ENST00000398250.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
ENST00000398250.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
ENST00000398250.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
ENST00000398250.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
ENST00000398250.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
ENST00000398250.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
ENST00000398250.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
ENST00000398250.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
ENST00000398250.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
ENST00000398250.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
ENST00000398250.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
ENST00000398250.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
ENST00000398250.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
ENST00000398250.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
ENST00000427079.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
ENST00000427079.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
ENST00000427079.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
ENST00000427079.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
ENST00000427079.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
ENST00000427079.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
ENST00000427079.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
ENST00000427079.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
ENST00000427079.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
ENST00000427079.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
ENST00000496437.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
ENST00000496437.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
NM_001030059.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
NM_001030059.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
NM_001030059.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
NM_001030059.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
NM_001030059.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
NM_001030059.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
NM_001030059.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
NM_001030059.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
NM_001030059.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
NM_001030059.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
NM_001030059.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
NM_001030059.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
NM_001030059.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
NM_001030059.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
NM_001030059.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009104.hg.1 |
NM_001030059.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10010587.hg.1 |
ENST00000369073.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10010587.hg.1 |
ENST00000398250.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10010587.hg.1 |
ENST00000541332.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10010587.hg.1 |
NM_001030059.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10010587.hg.1 |
TC10000874.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1000162299.hg.1 |
ENST00000369073.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000162299.hg.1 |
ENST00000398250.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000162299.hg.1 |
ENST00000427079.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000162299.hg.1 |
NM_001030059.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000162299.hg.1 |
ENST00000369073.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000162299.hg.1 |
ENST00000398250.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000162299.hg.1 |
ENST00000427079.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000162299.hg.1 |
NM_001030059.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000162299.hg.1 |
TC1000009104.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
ENST00000369073.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
ENST00000369073.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
ENST00000369073.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
ENST00000369073.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
ENST00000369073.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
ENST00000369073.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
ENST00000369073.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
ENST00000369073.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
ENST00000369073.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
ENST00000369073.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
ENST00000369073.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
ENST00000398250.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
ENST00000398250.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
ENST00000398250.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
ENST00000398250.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
ENST00000398250.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
ENST00000398250.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
ENST00000398250.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
ENST00000398250.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
ENST00000398250.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
ENST00000398250.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
ENST00000398250.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
ENST00000398250.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
ENST00000398250.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
ENST00000398250.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
ENST00000398250.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
ENST00000398250.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
ENST00000427079.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
ENST00000427079.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
ENST00000427079.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
ENST00000427079.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
ENST00000427079.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
ENST00000427079.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
ENST00000427079.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
ENST00000427079.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
ENST00000427079.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
ENST00000427079.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
ENST00000496437.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
ENST00000496437.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
NM_001030059.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
NM_001030059.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
NM_001030059.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
NM_001030059.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
NM_001030059.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
NM_001030059.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
NM_001030059.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
NM_001030059.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
NM_001030059.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
NM_001030059.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
NM_001030059.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
NM_001030059.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
NM_001030059.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
NM_001030059.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
NM_001030059.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009104.hg.1 |
NM_001030059.2 |
Refseq |