Assay Details
| Gene Symbol: RPL18A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK297323.1 | - | 5 | 978 | 74 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000222247.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000222247.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000222247.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000222247.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000222247.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000222247.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000222247.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000222247.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000222247.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000222247.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000384437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000597648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000597648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000597648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000597648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000597648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000597648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000597648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000597648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000597648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000599870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000599870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000599870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000599870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000599870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000599870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000599870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000599870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000599870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000599870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000599898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000599898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000599898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000599898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000599898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000599898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000599898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000600147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000600147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000600147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000600147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000600147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000600147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000600147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000600147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000600147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000600238.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000600238.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000600238.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000600238.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000600238.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000600238.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000600238.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000602216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000602216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000602216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000602216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000602216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | ENST00000602216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | NM_000980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | NM_000980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | NM_000980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | NM_000980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | NM_000980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | NM_000980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | NM_000980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | NM_000980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | NM_000980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | NM_000980.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007349.hg.1 | NR_000012.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000222247.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000222247.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000222247.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000222247.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000222247.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000222247.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000222247.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000222247.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000222247.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000222247.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000384437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000597648.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000597648.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000597648.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000597648.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000597648.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000597648.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000597648.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000597648.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000597648.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000599870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000599870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000599870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000599870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000599870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000599870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000599870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000599870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000599870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000599870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000599898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000599898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000599898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000599898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000599898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000599898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000599898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000600147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000600147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000600147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000600147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000600147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000600147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000600147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000600147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000600147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000600238.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000600238.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000600238.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000600238.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000600238.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000600238.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000600238.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000602216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000602216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000602216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000602216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000602216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | ENST00000602216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | NM_000980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | NM_000980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | NM_000980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | NM_000980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | NM_000980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | NM_000980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | NM_000980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | NM_000980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | NM_000980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | NM_000980.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007349.hg.1 | NR_000012.1 | Refseq |

RPL18A
Genomic Map