Assay Details
| Gene Symbol: PRSS57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214710.4 | NP_999875.1 | 1 - 2 | 113 | 102 | |
| GenBank mRNA | AY358594.1 | - | 1 - 2 | 150 | 102 | |
| BC137161.1 | - | 1 - 2 | 115 | 9021158 | 102 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000989.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19013491.hg.1 | ENST00000329267.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19013491.hg.1 | NM_214710.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19013491.hg.1 | TC19000989.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009104.hg.1 | ENST00000329267.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009104.hg.1 | ENST00000329267.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009104.hg.1 | ENST00000329267.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009104.hg.1 | ENST00000329267.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009104.hg.1 | ENST00000329267.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009104.hg.1 | ENST00000329267.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009104.hg.1 | ENST00000329267.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009104.hg.1 | ENST00000329267.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009104.hg.1 | ENST00000329267.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009104.hg.1 | ENST00000613411.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009104.hg.1 | ENST00000613411.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009104.hg.1 | ENST00000613411.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009104.hg.1 | ENST00000613411.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009104.hg.1 | ENST00000613411.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009104.hg.1 | ENST00000613411.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009104.hg.1 | ENST00000613411.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009104.hg.1 | ENST00000613411.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009104.hg.1 | ENST00000613411.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009104.hg.1 | ENST00000613411.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009104.hg.1 | ENST00000613411.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009104.hg.1 | NM_001308209.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009104.hg.1 | NM_001308209.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009104.hg.1 | NM_001308209.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009104.hg.1 | NM_001308209.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009104.hg.1 | NM_001308209.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009104.hg.1 | NM_001308209.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009104.hg.1 | NM_001308209.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009104.hg.1 | NM_001308209.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009104.hg.1 | NM_001308209.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009104.hg.1 | NM_001308209.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009104.hg.1 | NM_001308209.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009104.hg.1 | NM_214710.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009104.hg.1 | NM_214710.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009104.hg.1 | NM_214710.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009104.hg.1 | NM_214710.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009104.hg.1 | NM_214710.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009104.hg.1 | NM_214710.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009104.hg.1 | NM_214710.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009104.hg.1 | NM_214710.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009104.hg.1 | NM_214710.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009104.hg.1 | NM_214710.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009104.hg.1 | NM_214710.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19013493.hg.1 | ENST00000329267.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19013493.hg.1 | NM_214710.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19013493.hg.1 | TC19000989.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1900172312.hg.1 | ENST00000329267.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172312.hg.1 | ENST00000613411.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172312.hg.1 | NM_001308209.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900172312.hg.1 | NM_214710.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900172312.hg.1 | TC1900009104.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900172312.hg.1 | ENST00000329267.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172312.hg.1 | ENST00000613411.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172312.hg.1 | NM_001308209.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900172312.hg.1 | NM_214710.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009104.hg.1 | ENST00000329267.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009104.hg.1 | ENST00000329267.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009104.hg.1 | ENST00000329267.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009104.hg.1 | ENST00000329267.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009104.hg.1 | ENST00000329267.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009104.hg.1 | ENST00000329267.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009104.hg.1 | ENST00000329267.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009104.hg.1 | ENST00000329267.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009104.hg.1 | ENST00000329267.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009104.hg.1 | ENST00000613411.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009104.hg.1 | ENST00000613411.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009104.hg.1 | ENST00000613411.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009104.hg.1 | ENST00000613411.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009104.hg.1 | ENST00000613411.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009104.hg.1 | ENST00000613411.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009104.hg.1 | ENST00000613411.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009104.hg.1 | ENST00000613411.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009104.hg.1 | ENST00000613411.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009104.hg.1 | ENST00000613411.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009104.hg.1 | ENST00000613411.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009104.hg.1 | NM_001308209.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009104.hg.1 | NM_001308209.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009104.hg.1 | NM_001308209.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009104.hg.1 | NM_001308209.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009104.hg.1 | NM_001308209.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009104.hg.1 | NM_001308209.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009104.hg.1 | NM_001308209.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009104.hg.1 | NM_001308209.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009104.hg.1 | NM_001308209.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009104.hg.1 | NM_001308209.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009104.hg.1 | NM_001308209.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009104.hg.1 | NM_214710.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009104.hg.1 | NM_214710.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009104.hg.1 | NM_214710.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009104.hg.1 | NM_214710.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009104.hg.1 | NM_214710.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009104.hg.1 | NM_214710.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009104.hg.1 | NM_214710.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009104.hg.1 | NM_214710.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009104.hg.1 | NM_214710.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009104.hg.1 | NM_214710.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009104.hg.1 | NM_214710.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900172315.hg.1 | ENST00000329267.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172315.hg.1 | ENST00000613411.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172315.hg.1 | NM_001308209.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900172315.hg.1 | NM_214710.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900172315.hg.1 | TC1900009104.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900172315.hg.1 | ENST00000329267.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172315.hg.1 | ENST00000613411.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172315.hg.1 | NM_001308209.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900172315.hg.1 | NM_214710.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900172313.hg.1 | ENST00000329267.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172313.hg.1 | ENST00000613411.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172313.hg.1 | NM_001308209.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900172313.hg.1 | NM_214710.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900172313.hg.1 | TC1900009104.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900172313.hg.1 | ENST00000329267.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172313.hg.1 | ENST00000613411.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172313.hg.1 | NM_001308209.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900172313.hg.1 | NM_214710.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900172316.hg.1 | ENST00000329267.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172316.hg.1 | ENST00000613411.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172316.hg.1 | NM_001308209.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900172316.hg.1 | NM_214710.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900172316.hg.1 | TC1900009104.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900172316.hg.1 | ENST00000329267.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172316.hg.1 | ENST00000613411.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172316.hg.1 | NM_001308209.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900172316.hg.1 | NM_214710.4 | Refseq |

PRSS57
Genomic Map