Assay Details
| Gene Symbol: COL1A1-PDGFB fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | Y15916.1 | - | 150 | 136 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

COL1A1-PDGFB fusion| Gene Symbol: COL1A1-PDGFB fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | Y15916.1 | - | 150 | 136 | ||
Set Membership: |
|