Assay Details
| Gene Symbol: MRPL52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181307.2 | NP_851824.1 | 2 - 3 | 121 | 125 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14000132.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR14001117.hg.1 | ENST00000355151.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001117.hg.1 | ENST00000397496.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001117.hg.1 | NM_178336.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001117.hg.1 | NM_180982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001117.hg.1 | NM_181304.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001117.hg.1 | NM_181305.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001117.hg.1 | NM_181306.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001117.hg.1 | NM_181307.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001117.hg.1 | TC14000132.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001114.hg.1 | ENST00000311892.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001114.hg.1 | ENST00000355151.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001114.hg.1 | ENST00000397496.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001114.hg.1 | ENST00000397505.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001114.hg.1 | ENST00000432849.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001114.hg.1 | ENST00000553711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001114.hg.1 | ENST00000555345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001114.hg.1 | ENST00000555536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001114.hg.1 | ENST00000556840.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001114.hg.1 | ENST00000557221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001114.hg.1 | NM_178336.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001114.hg.1 | NM_180982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001114.hg.1 | NM_181304.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001114.hg.1 | NM_181305.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001114.hg.1 | NM_181306.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001114.hg.1 | NM_181307.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001114.hg.1 | TC14000132.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001116.hg.1 | ENST00000355151.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001116.hg.1 | NM_178336.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001116.hg.1 | NM_180982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001116.hg.1 | NM_181304.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001116.hg.1 | NM_181305.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001116.hg.1 | NM_181306.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001116.hg.1 | NM_181307.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001116.hg.1 | TC14000132.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001120.hg.1 | ENST00000355151.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001120.hg.1 | ENST00000397496.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001120.hg.1 | ENST00000432849.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001120.hg.1 | ENST00000553711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001120.hg.1 | ENST00000555345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001120.hg.1 | NM_178336.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001120.hg.1 | NM_180982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001120.hg.1 | NM_181304.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001120.hg.1 | NM_181305.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001120.hg.1 | NM_181306.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001120.hg.1 | NM_181307.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001120.hg.1 | TC14000132.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001121.hg.1 | ENST00000355151.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001121.hg.1 | ENST00000397496.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001121.hg.1 | ENST00000432849.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001121.hg.1 | ENST00000553711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001121.hg.1 | ENST00000555345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001121.hg.1 | NM_178336.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001121.hg.1 | NM_180982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001121.hg.1 | NM_181304.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001121.hg.1 | NM_181305.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001121.hg.1 | NM_181306.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001121.hg.1 | NM_181307.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001121.hg.1 | TC14000132.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001118.hg.1 | ENST00000355151.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001118.hg.1 | ENST00000397496.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001118.hg.1 | NM_178336.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001118.hg.1 | NM_180982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001118.hg.1 | NM_181304.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001118.hg.1 | NM_181305.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001118.hg.1 | NM_181306.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001118.hg.1 | NM_181307.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001118.hg.1 | TC14000132.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001119.hg.1 | ENST00000355151.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001119.hg.1 | ENST00000397496.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001119.hg.1 | ENST00000432849.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001119.hg.1 | ENST00000555345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001119.hg.1 | NM_178336.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001119.hg.1 | NM_180982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001119.hg.1 | NM_181304.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001119.hg.1 | NM_181305.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001119.hg.1 | NM_181306.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001119.hg.1 | NM_181307.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001119.hg.1 | TC14000132.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001125.hg.1 | ENST00000311892.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001125.hg.1 | ENST00000355151.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001125.hg.1 | ENST00000397496.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001125.hg.1 | ENST00000397505.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001125.hg.1 | ENST00000432849.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001125.hg.1 | ENST00000553711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001125.hg.1 | ENST00000555345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001125.hg.1 | ENST00000555536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001125.hg.1 | ENST00000556840.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001125.hg.1 | ENST00000557221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001125.hg.1 | NM_178336.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001125.hg.1 | NM_180982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001125.hg.1 | NM_181304.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001125.hg.1 | NM_181305.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001125.hg.1 | NM_181306.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001125.hg.1 | NM_181307.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001125.hg.1 | TC14000132.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001122.hg.1 | ENST00000355151.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001122.hg.1 | ENST00000397496.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001122.hg.1 | ENST00000397505.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001122.hg.1 | ENST00000432849.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001122.hg.1 | ENST00000553711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001122.hg.1 | ENST00000555345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001122.hg.1 | NM_178336.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001122.hg.1 | NM_180982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001122.hg.1 | NM_181304.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001122.hg.1 | NM_181305.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001122.hg.1 | NM_181306.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001122.hg.1 | NM_181307.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14001122.hg.1 | TC14000132.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1400145491.hg.1 | ENST00000311892.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145491.hg.1 | ENST00000355151.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145491.hg.1 | ENST00000397496.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145491.hg.1 | ENST00000397505.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145491.hg.1 | ENST00000432849.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145491.hg.1 | ENST00000461594.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145491.hg.1 | ENST00000553482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145491.hg.1 | ENST00000553711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145491.hg.1 | ENST00000555345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145491.hg.1 | ENST00000555536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145491.hg.1 | ENST00000556214.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145491.hg.1 | ENST00000556465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145491.hg.1 | ENST00000556654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145491.hg.1 | ENST00000556840.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145491.hg.1 | ENST00000557221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145491.hg.1 | ENST00000557543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145491.hg.1 | NM_178336.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400145491.hg.1 | NM_180982.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400145491.hg.1 | NM_181304.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400145491.hg.1 | NM_181305.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400145491.hg.1 | NM_181306.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400145491.hg.1 | NM_181307.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400145491.hg.1 | ENST00000311892.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145491.hg.1 | ENST00000355151.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145491.hg.1 | ENST00000397496.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145491.hg.1 | ENST00000397505.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145491.hg.1 | ENST00000432849.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145491.hg.1 | ENST00000461594.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145491.hg.1 | ENST00000553482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145491.hg.1 | ENST00000553711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145491.hg.1 | ENST00000555345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145491.hg.1 | ENST00000555536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145491.hg.1 | ENST00000556214.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145491.hg.1 | ENST00000556465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145491.hg.1 | ENST00000556654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145491.hg.1 | ENST00000556840.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145491.hg.1 | ENST00000557221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145491.hg.1 | ENST00000557543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145491.hg.1 | NM_178336.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400145491.hg.1 | NM_180982.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400145491.hg.1 | NM_181304.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400145491.hg.1 | NM_181305.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400145491.hg.1 | NM_181306.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400145491.hg.1 | NM_181307.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400145491.hg.1 | TC1400006658.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400145490.hg.1 | ENST00000311892.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145490.hg.1 | ENST00000355151.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145490.hg.1 | ENST00000397496.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145490.hg.1 | ENST00000397505.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145490.hg.1 | ENST00000432849.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145490.hg.1 | ENST00000461594.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145490.hg.1 | ENST00000553482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145490.hg.1 | ENST00000553711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145490.hg.1 | ENST00000555345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145490.hg.1 | ENST00000555536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145490.hg.1 | ENST00000556214.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145490.hg.1 | ENST00000556465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145490.hg.1 | ENST00000556840.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145490.hg.1 | ENST00000557221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145490.hg.1 | ENST00000557543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145490.hg.1 | NM_178336.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400145490.hg.1 | NM_180982.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400145490.hg.1 | NM_181304.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400145490.hg.1 | NM_181305.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400145490.hg.1 | NM_181306.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400145490.hg.1 | NM_181307.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400145490.hg.1 | ENST00000311892.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145490.hg.1 | ENST00000355151.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145490.hg.1 | ENST00000397496.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145490.hg.1 | ENST00000397505.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145490.hg.1 | ENST00000432849.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145490.hg.1 | ENST00000461594.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145490.hg.1 | ENST00000553482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145490.hg.1 | ENST00000553711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145490.hg.1 | ENST00000555345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145490.hg.1 | ENST00000555536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145490.hg.1 | ENST00000556214.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145490.hg.1 | ENST00000556465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145490.hg.1 | ENST00000556840.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145490.hg.1 | ENST00000557221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145490.hg.1 | ENST00000557543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145490.hg.1 | NM_178336.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400145490.hg.1 | NM_180982.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400145490.hg.1 | NM_181304.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400145490.hg.1 | NM_181305.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400145490.hg.1 | NM_181306.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400145490.hg.1 | NM_181307.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400145490.hg.1 | TC1400006658.hg.1 | Clariom D TC |

MRPL52
Genomic Map