Assay Details
| Gene Symbol: SNHG15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003697.1 | 1 - 2 | 139 | 78 | ||
| GenBank mRNA | BC092459.1 | - | 1 - 2 | 139 | 78 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07001349.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000384215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000384215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000438705.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000438705.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000438705.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000438705.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000438705.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000438705.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000577700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000577700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000577700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000577700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000577700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000577700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000577700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000577700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000577700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000577700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000577700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000577700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000577700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000577700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000577700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000577700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000577700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000577700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000577700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000577700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000577700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000577700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000577700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000577700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000577700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000577700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000577700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000577700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000577700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000577700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000577700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000577700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000577700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000577700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000578968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000578968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000578968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000578968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000578968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000578968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000578968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000578968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000578968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000578968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000578968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000578968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000578968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000578968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000578968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000578968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000578968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000578968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000578968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000578968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000578968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000578968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000579383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000579383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000579383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000579383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000579383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000579383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000579383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000579383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000579383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000579383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000579383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000579383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000579383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000579383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000579383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000579383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000579383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000579383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000580458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000580458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000580458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000580458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000580458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000580458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000580458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000580458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000580458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000580458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000580458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000580458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000580458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000580458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000580458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000580458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000580458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000580458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000580528.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000580528.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000580528.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000580528.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000580528.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000580528.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000580528.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000580528.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000582727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000582727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000582727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000582727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000582727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000582727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000582727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000582727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000582727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000584327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000584327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000584327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000584327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000584327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000584327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000584327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000584327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000584327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000584327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000584327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000584327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000584686.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000584686.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000584686.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000584686.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000584686.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000584686.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000585030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000585030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000585030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000585030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000585030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000585030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000585030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000585030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000585030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000585030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000585030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000585030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000585030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000585030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000585030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000585030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000585030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000585030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000585030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000585030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000585030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | ENST00000585030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | NR_002952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | NR_002952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | NR_003697.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | NR_003697.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | NR_003697.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | NR_003697.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | NR_003697.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | NR_003697.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | NR_003697.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | NR_003697.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | NR_003697.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010942.hg.1 | NR_003697.1 | Refseq |

SNHG15
Genomic Map