Assay Details
| Gene Symbol: EMX2OS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002791.2 | 1 - 2 | 497 | 72 | ||
| GenBank mRNA | AY117034.1 | - | 1 - 2 | 497 | 72 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | PDZ domain containing 8 | PDZD8 | PDZK8 | -168.8kb | Sense | Chr. 10 - 117280489 - 117375467 on NCBI Build GRCh38 |
| Downstream | RAB11 family interacting protein 2 | RAB11FIP2 |
Rab11-FIP2, nRip11 |
459.8kb | Sense | Chr. 10 - 118004916 - 118046921 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001689.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10022247.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10022247.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10022247.hg.1 | TC10001689.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000423419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000423419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000423419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000423419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000424371.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000424371.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000424371.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000424371.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000450314.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000450314.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000450314.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000450314.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000450314.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000450314.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000450314.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000450314.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000611404.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000611404.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | ENST00000611404.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10022248.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10022248.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10022248.hg.1 | TC10001689.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1000181826.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000181826.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000181826.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000181826.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000181826.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000181826.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000181826.hg.1 | TC1000011951.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000181825.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000181825.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000181825.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000181825.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000181825.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000181825.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000181825.hg.1 | TC1000011951.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000412075.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000423419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000423419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000423419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000423419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000424371.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000424371.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000424371.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000424371.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000440007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000450314.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000450314.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000450314.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000450314.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000450314.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000450314.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000450314.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000450314.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000551288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000611404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000611404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | ENST00000611404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | NR_002791.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011951.hg.1 | NR_002791.2 | Refseq |

EMX2OS
Genomic Map