Assay Details
| Gene Symbol: LOC729187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK130923.1 | - | 1 | 1254 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | KCNMA1 antisense RNA 3 | KCNMA1-AS3 | -78.5kb | Anti-sense | Chr. 10 - 77350872 - 77376613 on NCBI Build GRCh38 | |
| Downstream | discs large MAGUK scaffold protein 5 | DLG5 |
LP-DLG, P-DLG5, PDLG |
334.1kb | Sense | Chr. 10 - 77790787 - 77926590 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC729187
Genomic Map