Assay Details
Gene Symbol: GKN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AL833013.1 | - | 1 | 2044 | 102 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | gastrokine 2 | GKN2 |
BRICD1B, GDDR, PRO813, TFIZ1, VLTI465 |
-25.6kb | Anti-sense | Chr. 2 - 68945232 - 68952970 on NCBI Build GRCh38 |
Downstream | anthrax toxin receptor 1 | ANTXR1 |
ATR, GAPO, TEM8 |
32.2kb | Sense | Chr. 2 - 69013144 - 69249327 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
