Assay Details
| Gene Symbol: LOC100129408 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK127916.1 | - | 1 | 147 | 79 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC100505549 | LOC100505549 | Sense | Chr. 18 - 57630302 - 57669276 on NCBI Build GRCh38 | ||
| Downstream |
neural precursor cell expressed, developmentally down-regulated 4-like, E3 ubiquitin protein ligase |
NEDD4L |
NEDD4-2, NEDD4.2, RSP5, hNEDD4-2 |
384.6kb | Sense | Chr. 18 - 58044362 - 58401540 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100129408
Genomic Map