Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | PHKA2 antisense RNA 1 | PHKA2-AS1 | -4.1kb | Sense | Chr. X - 18890296 - 18894975 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC101928415 | LOC101928415 | 83.0kb | Sense | Chr. X - 18984254 - 19082825 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map