Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
RNA polymerase II subunit J4, pseudogene |
POLR2J4 | RPB11-phi | Sense | Chr. 7 - 43940895 - 44019194 on NCBI Build GRCh38 | |
| Downstream | speedy/RINGO cell cycle regulator family member E1 | SPDYE1 |
Ringo1, SPDYE, WBSCR19 |
36.0kb | Anti-sense | Chr. 7 - 43997855 - 44011040 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map