Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | KCNMA1 antisense RNA 2 | KCNMA1-AS2 | -103.0kb | Sense | Chr. 10 - 77140831 - 77150100 on NCBI Build GRCh38 | |
| Downstream | KCNMA1 antisense RNA 3 | KCNMA1-AS3 | 95.7kb | Sense | Chr. 10 - 77350872 - 77376613 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map