Assay Details
| Gene Symbol: PTPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK097317.1 | - | 1 | 1356 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | carnitine O-acetyltransferase | CRAT | CAT1 | -32.3kb | Anti-sense | Chr. 9 - 129094794 - 129110791 on NCBI Build GRCh38 |
| Downstream | immediate early response 5 like | IER5L | bA247A12.2 | 30.8kb | Anti-sense | Chr. 9 - 129175552 - 129178261 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

PTPA
Genomic Map