Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC100507283 | LOC100507283 | Sense | Chr. 11 - 124800428 - 124834487 on NCBI Build GRCh38 | ||
| Downstream | roundabout guidance receptor 3 | ROBO3 |
HGPPS, HGPS, RBIG1, RIG1 |
55.6kb | Sense | Chr. 11 - 124865409 - 124881474 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map