Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 4445 | MIR4445 | -1.3mb | Sense | Chr. 3 - 109602828 - 109602897 on NCBI Build GRCh38 | |
| Downstream | PVRL3 antisense RNA 1 | PVRL3-AS1 | 123.7kb | Anti-sense | Chr. 3 - 111045315 - 111069959 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map