Assay Details
| Gene Symbol: POLRMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005035.3 | NP_005026.3 | 1 - 2 | 145 | 92 | |
| XM_005259580.3 | XP_005259637.2 | 1 - 2 | 169 | 92 | ||
| GenBank mRNA | AK225723.1 | - | 1 - 2 | 106 | 92 | |
| AK225865.1 | - | 1 - 2 | 106 | 92 | ||
| AK302877.1 | - | 1 - 2 | 144 | 92 | ||
| AK307525.1 | - | 1 - 2 | 90 | 92 | ||
| BC030829.1 | - | 1 - 2 | 98 | 92 | ||
| BC098387.1 | - | 1 - 2 | 126 | 5264127 | 92 | |
| U75370.1 | - | 1 - 2 | 121 | 92 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000987.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19013461.hg.1 | ENST00000215591.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19013461.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19013461.hg.1 | TC19000987.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000587057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000587057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000587057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000587057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000587057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000587057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000587057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000587057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000587057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000587057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000589961.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000589961.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000589961.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000589961.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000589961.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000590336.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000590336.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000590336.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000590336.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000590336.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000590573.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000590573.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000590573.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000590573.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000590573.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000590709.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000590709.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000590709.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000590709.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000590709.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000590709.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000590709.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000590709.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000592633.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000592633.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000592633.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000592633.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000592633.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000592633.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000592863.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000592863.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000592863.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | ENST00000592863.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000587057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000587057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000587057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000587057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000587057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000587057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000587057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000587057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000587057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000587057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000589961.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000589961.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000589961.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000589961.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000589961.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000590336.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000590336.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000590336.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000590336.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000590336.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000590573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000590573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000590573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000590573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000590573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000590709.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000590709.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000590709.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000590709.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000590709.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000590709.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000590709.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000590709.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000592633.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000592633.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000592633.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000592633.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000592633.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000592633.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000592863.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000592863.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000592863.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | ENST00000592863.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009100.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900172253.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172253.hg.1 | ENST00000590573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172253.hg.1 | ENST00000592863.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172253.hg.1 | NM_005035.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900172253.hg.1 | TC1900009100.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900172253.hg.1 | ENST00000588649.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172253.hg.1 | ENST00000590573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172253.hg.1 | ENST00000592863.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900172253.hg.1 | NM_005035.3 | Refseq |

POLRMT
Genomic Map