Assay Details
| Gene Symbol: PVRL3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045114.1 | 1 - 2 | 165 | 148 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | putative uncharacterized protein LOC151760 | LOC151760 | -98.3kb | Sense | Chr. 3 - 110892123 - 110974348 on NCBI Build GRCh38 | |
| Downstream | nectin cell adhesion molecule 3 | NECTIN3 |
CD113, CDW113, NECTIN-3, PPR3, PRR3, PVRL3, PVRR3 |
1.8kb | Anti-sense | Chr. 3 - 111071759 - 111201444 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001642.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03029626.hg.1 | ENST00000463025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029626.hg.1 | ENST00000467426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029626.hg.1 | ENST00000474769.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029626.hg.1 | ENST00000476301.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029626.hg.1 | ENST00000491709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029626.hg.1 | NR_045114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029626.hg.1 | TC03001642.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029627.hg.1 | ENST00000467426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029627.hg.1 | ENST00000474769.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029627.hg.1 | ENST00000476301.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029627.hg.1 | ENST00000491709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029627.hg.1 | NR_045114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029627.hg.1 | TC03001642.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029628.hg.1 | ENST00000467426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029628.hg.1 | NR_045114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029628.hg.1 | TC03001642.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | ENST00000463025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | ENST00000463025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | ENST00000463025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | ENST00000467426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | ENST00000467426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | ENST00000467426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | ENST00000467426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | ENST00000467426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | ENST00000467426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | ENST00000467426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | ENST00000467426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | ENST00000467426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | ENST00000467426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | ENST00000474769.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | ENST00000474769.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | ENST00000474769.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | ENST00000474769.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | ENST00000474769.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | ENST00000474769.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | ENST00000474769.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | ENST00000474769.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | ENST00000474769.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | ENST00000474769.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | ENST00000476301.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | ENST00000476301.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | ENST00000476301.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | ENST00000476301.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | ENST00000476301.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | ENST00000476301.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | ENST00000476301.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | ENST00000476301.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | ENST00000476301.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | ENST00000476301.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | ENST00000491709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | ENST00000491709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | ENST00000491709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | ENST00000491709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | ENST00000491709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | ENST00000491709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | ENST00000491709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | ENST00000491709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | ENST00000491709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | ENST00000491709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | ENST00000491709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | ENST00000491709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | NR_045114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | NR_045114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | NR_045114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | NR_045114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | NR_045114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | NR_045114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011974.hg.1 | NR_045114.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300189125.hg.1 | ENST00000467426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189125.hg.1 | ENST00000467426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189125.hg.1 | NR_045114.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300189125.hg.1 | ENST00000467426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189125.hg.1 | ENST00000467426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189125.hg.1 | NR_045114.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300189125.hg.1 | TC0300011974.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300189123.hg.1 | ENST00000463025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189123.hg.1 | ENST00000467426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189123.hg.1 | ENST00000467426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189123.hg.1 | ENST00000474769.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189123.hg.1 | ENST00000474769.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189123.hg.1 | ENST00000476301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189123.hg.1 | ENST00000476301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189123.hg.1 | ENST00000491709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189123.hg.1 | ENST00000491709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189123.hg.1 | NR_045114.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300189123.hg.1 | ENST00000463025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189123.hg.1 | ENST00000467426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189123.hg.1 | ENST00000467426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189123.hg.1 | ENST00000474769.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189123.hg.1 | ENST00000474769.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189123.hg.1 | ENST00000476301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189123.hg.1 | ENST00000476301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189123.hg.1 | ENST00000491709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189123.hg.1 | ENST00000491709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189123.hg.1 | NR_045114.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300189123.hg.1 | TC0300011974.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300189124.hg.1 | ENST00000467426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189124.hg.1 | ENST00000467426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189124.hg.1 | ENST00000474769.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189124.hg.1 | ENST00000474769.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189124.hg.1 | ENST00000476301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189124.hg.1 | ENST00000476301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189124.hg.1 | ENST00000491709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189124.hg.1 | ENST00000491709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189124.hg.1 | NR_045114.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300189124.hg.1 | ENST00000467426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189124.hg.1 | ENST00000467426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189124.hg.1 | ENST00000474769.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189124.hg.1 | ENST00000474769.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189124.hg.1 | ENST00000476301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189124.hg.1 | ENST00000476301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189124.hg.1 | ENST00000491709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189124.hg.1 | ENST00000491709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189124.hg.1 | NR_045114.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300189124.hg.1 | TC0300011974.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | ENST00000463025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | ENST00000463025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | ENST00000463025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | ENST00000467426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | ENST00000467426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | ENST00000467426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | ENST00000467426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | ENST00000467426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | ENST00000467426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | ENST00000467426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | ENST00000467426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | ENST00000467426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | ENST00000467426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | ENST00000474769.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | ENST00000474769.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | ENST00000474769.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | ENST00000474769.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | ENST00000474769.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | ENST00000474769.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | ENST00000474769.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | ENST00000474769.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | ENST00000474769.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | ENST00000474769.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | ENST00000476301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | ENST00000476301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | ENST00000476301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | ENST00000476301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | ENST00000476301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | ENST00000476301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | ENST00000476301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | ENST00000476301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | ENST00000476301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | ENST00000476301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | ENST00000491709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | ENST00000491709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | ENST00000491709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | ENST00000491709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | ENST00000491709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | ENST00000491709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | ENST00000491709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | ENST00000491709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | ENST00000491709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | ENST00000491709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | ENST00000491709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | ENST00000491709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | NR_045114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | NR_045114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | NR_045114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | NR_045114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | NR_045114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | NR_045114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011974.hg.1 | NR_045114.1 | Refseq |

PVRL3-AS1
Genomic Map