Assay Details
| Gene Symbol: USP2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_034160.1 | 1 - 2 | 136 | 88 | ||
| GenBank mRNA | AK097820.1 | - | 1 - 2 | 104 | 88 | |
| AL559224.3 | - | 1 - 2 | 89 | 88 | ||
| BQ429648.1 | - | 1 - 2 | 139 | 88 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000498979.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000498979.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000498979.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000498979.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000498979.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000498979.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000498979.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000498979.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000498979.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000498979.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000498979.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000498979.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000500970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000500970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000500970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000500970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000500970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000500970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000500970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000500970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000500970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000500970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000500970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000500970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000500970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000500970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000500970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000500970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000500970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000500970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000500970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000500970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000530002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000530002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000530002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000530002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000530002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000577297.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000577297.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000577297.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000577297.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000577297.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000577297.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000577297.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000577297.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000577297.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000577297.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000577297.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000577297.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000578923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000578923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | ENST00000578923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | NR_034160.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | NR_034160.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | NR_034160.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | NR_034160.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | NR_034160.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013105.hg.1 | NR_034160.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000498979.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000498979.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000498979.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000498979.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000498979.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000498979.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000498979.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000498979.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000498979.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000498979.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000498979.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000498979.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000500970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000500970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000500970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000500970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000500970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000500970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000500970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000500970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000500970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000500970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000500970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000500970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000500970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000500970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000500970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000500970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000500970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000500970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000500970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000500970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000530002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000530002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000530002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000530002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000530002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000577297.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000577297.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000577297.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000577297.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000577297.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000577297.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000577297.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000577297.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000577297.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000577297.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000577297.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000577297.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000578923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000578923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | ENST00000578923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | NR_034160.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | NR_034160.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | NR_034160.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | NR_034160.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | NR_034160.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013105.hg.1 | NR_034160.1 | Refseq |

USP2-AS1
Genomic Map