Assay Details
| Gene Symbol: LOC100287290 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY070437.1 | - | 1 - 2 | 47 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AY070437.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000465477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000465477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000465477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000465477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000465477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000465477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000465477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000465477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000465477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000465477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000465477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000465477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000465477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000465477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000468242.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000468242.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000468242.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000468242.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000468242.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000468242.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000468242.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000468242.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000468242.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000468242.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000468242.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000468242.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000468242.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000468242.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000468242.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000468242.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000477589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000477589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000477589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000477589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000477589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000477589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000477589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000477589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000477589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000477589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000477589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000477589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000495318.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000495318.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000495318.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000495318.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000495318.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000495318.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000495318.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000495318.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000495318.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000495318.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000495318.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000495318.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000496842.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000496842.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000496842.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000607426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000607426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013895.hg.1 | ENST00000607426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000465477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000465477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000465477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000465477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000465477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000465477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000465477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000465477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000465477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000465477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000465477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000465477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000465477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000465477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000468242.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000468242.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000468242.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000468242.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000468242.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000468242.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000468242.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000468242.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000468242.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000468242.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000468242.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000468242.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000468242.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000468242.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000468242.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000468242.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000477589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000477589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000477589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000477589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000477589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000477589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000477589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000477589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000477589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000477589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000477589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000477589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000495318.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000495318.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000495318.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000495318.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000495318.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000495318.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000495318.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000495318.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000495318.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000495318.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000495318.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000495318.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000496842.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000496842.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000496842.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000607426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000607426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013895.hg.1 | ENST00000607426.1 | Ensembl |

LOC100287290
Genomic Map