Assay Details
| Gene Symbol: PLPP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318169.1 | NP_001305098.1 | 2 - 3 | 521 | 99 | |
| GenBank mRNA | BC122535.1 | - | 3 - 4 | 318 | 99 | |
More Information
Set Membership: |
|

PLPP4
Genomic Map