Assay Details
| Gene Symbol: ASRGL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083926.1 | NP_001077395.1 | 7 | 1952 | 94 | |
| NM_025080.3 | NP_079356.3 | 7 | 1875 | 94 | ||
| XM_005274305.3 | XP_005274362.1 | 6 | 1592 | 94 | ||
| XM_005274306.2 | XP_005274363.1 | 6 | 1654 | 94 | ||
| GenBank mRNA | AK090908.1 | - | 7 | 1724 | 94 | |
| AK091894.1 | - | 7 | 1800 | 94 | ||
| BC006267.2 | - | 4 | 1311 | 94 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000301776.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000301776.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000301776.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000301776.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000301776.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000301776.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000301776.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000301776.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000301776.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000301776.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000301776.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000525496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000525496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000525496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000525496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000525708.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000525708.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000525708.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000526096.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000526096.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000526096.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000526096.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000526096.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000528206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000528206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000529226.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000529226.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000529226.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000529226.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000529226.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000529226.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000533970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000533970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000533970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000534183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000534183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000534183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000534183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000534183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000534183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000534183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000534571.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000534571.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000534571.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000534571.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000534571.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000534571.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000628829.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000628829.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000628829.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000628829.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000628829.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000628829.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000628829.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000628829.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | ENST00000628829.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011078.hg.1 | ENST00000526045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011078.hg.1 | ENST00000526045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000301776.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000301776.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000301776.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000301776.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000301776.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000301776.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000301776.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000301776.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000301776.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000301776.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000301776.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000415229.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000525496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000525496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000525496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000525496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000525708.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000525708.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000525708.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000526096.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000526096.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000526096.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000526096.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000526096.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000528206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000528206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000529226.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000529226.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000529226.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000529226.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000529226.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000529226.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000533970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000533970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000533970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000534183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000534183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000534183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000534183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000534183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000534183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000534183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000534571.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000534571.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000534571.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000534571.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000534571.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000534571.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000628829.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000628829.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000628829.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000628829.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000628829.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000628829.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000628829.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000628829.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | ENST00000628829.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_001083926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007844.hg.1 | NM_025080.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011078.hg.1 | ENST00000526045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011078.hg.1 | ENST00000526045.1 | Ensembl |

ASRGL1
Genomic Map