Assay Details
| Gene Symbol: TBL1XR1-PIK3CA fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_TBL1XR1-PIK3CA_T1P2.0 | - | 35 | 88 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TBL1XR1-PIK3CA fusion| Gene Symbol: TBL1XR1-PIK3CA fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_TBL1XR1-PIK3CA_T1P2.0 | - | 35 | 88 | ||