Assay Details
| Gene Symbol: MPRIP-RAF1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_MPRIP-RAF1_M22R8.0 | - | 123 | 106 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MPRIP-RAF1 fusion| Gene Symbol: MPRIP-RAF1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_MPRIP-RAF1_M22R8.0 | - | 123 | 106 | ||
Set Membership: |
|