Assay Details
| Gene Symbol: TMEM106B-ROS1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_TMEM106B-ROS1_T3R35.0 | - | 119 | 97 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM106B-ROS1 fusion| Gene Symbol: TMEM106B-ROS1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_TMEM106B-ROS1_T3R35.0 | - | 119 | 97 | ||