Assay Details
| Gene Symbol: PCM1-JAK2 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_PCM1-JAK2_P26J9_COSF1007.0 | - | 74 | 121 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PCM1-JAK2 fusion| Gene Symbol: PCM1-JAK2 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_PCM1-JAK2_P26J9_COSF1007.0 | - | 74 | 121 | ||
Set Membership: |
|