Assay Details
| Gene Symbol: RNF11-BRAF fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_RNF11-BRAF_R1B11.0 | - | 71 | 89 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RNF11-BRAF fusion| Gene Symbol: RNF11-BRAF fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_RNF11-BRAF_R1B11.0 | - | 71 | 89 | ||