Assay Details
| Gene Symbol: PARK2-FGFR2 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_PARK2-FGFR2_P9F11.0 | - | 101 | 90 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PARK2-FGFR2 fusion| Gene Symbol: PARK2-FGFR2 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_PARK2-FGFR2_P9F11.0 | - | 101 | 90 | ||