Assay Details
| Gene Symbol: LINC00957 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015401.2 | 2 | 600 | 89 | ||
| NR_072981.1 | 2 | 600 | 89 | |||
| NR_072982.1 | 2 | 466 | 89 | |||
| GenBank mRNA | AK092709.1 | - | 2 | 542 | 89 | |
| BC081571.1 | - | 2 | 478 | 89 | ||
| DA204840.1 | - | 2 | 437 | 89 | ||
| DA293391.1 | - | 2 | 435 | 89 | ||
| DA406714.1 | - | 2 | 417 | 89 | ||
More Information
Set Membership: |
|

LINC00957
Genomic Map