Assay Details
| Gene Symbol: NDUFB8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284367.1 | NP_001271296.1 | 3 - 4 | 391 | 66 | |
| NM_001284368.1 | NP_001271297.1 | 3 - 4 | 300 | 66 | ||
| NM_005004.3 | NP_004995.1 | 3 - 4 | 391 | 66 | ||
| GenBank mRNA | AA782554.1 | - | 2 - 3 | 368 | 66 | |
| AF044958.1 | - | 3 - 4 | 341 | 66 | ||
| AF077028.1 | - | 3 - 4 | 338 | 66 | ||
| AF115968.1 | - | 3 - 4 | 347 | 66 | ||
| AK289579.1 | - | 3 - 4 | 332 | 66 | ||
| AL080056.1 | - | 3 - 4 | 351 | 66 | ||
| BC000466.2 | - | 3 - 4 | 332 | 2822781 | 66 | |
| BC019276.1 | - | 3 - 4 | 332 | 2822781 | 66 | |
| BM008944.1 | - | 3 - 4 | 288 | 66 | ||
| BM686677.1 | - | 3 - 4 | 339 | 66 | ||
| CF127298.1 | - | 3 - 4 | 330 | 66 | ||
| CR533490.1 | - | 3 - 4 | 319 | 66 | ||
| DA428446.1 | - | 3 - 4 | 391 | 66 | ||
More Information
Set Membership: |
|

NDUFB8
Genomic Map