Assay Details
| Gene Symbol: NDUFB8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284367.1 | NP_001271296.1 | 4 - 5 | 543 | 117 | |
| GenBank mRNA | BM686677.1 | - | 4 - 5 | 491 | 117 | |
More Information
Set Membership: |
|

NDUFB8
Genomic Map