Assay Details
| Gene Symbol: POLR2M | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001018102.2 | NP_001018112.1 | 1 - 2 | 439 | 61 | |
| GenBank mRNA | AK128618.1 | - | 1 - 2 | 235 | 61 | |
| AY353061.1 | - | 1 - 2 | 168 | 61 | ||
More Information
Set Membership: |
|

POLR2M
Genomic Map