Assay Details
| Gene Symbol: NDUFA12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258338.1 | NP_001245267.1 | 1 - 2 | 125 | 69 | |
| NM_018838.4 | NP_061326.1 | 1 - 2 | 125 | 69 | ||
| GenBank mRNA | AF112208.1 | - | 1 - 2 | 145 | 69 | |
| AF217092.1 | - | 1 - 2 | 104 | 69 | ||
| AK222619.1 | - | 1 - 2 | 109 | 69 | ||
| BC005936.1 | - | 1 - 2 | 114 | 4069306 | 69 | |
| BG943528.1 | - | 1 - 2 | 110 | 69 | ||
| BM548072.1 | - | 192 | 69 | |||
| BT007220.1 | - | 1 - 2 | 92 | 69 | ||
| BU532909.1 | - | 1 - 2 | 112 | 69 | ||
| DW441120.1 | - | 1 - 2 | 139 | 69 | ||
More Information
Set Membership: |
|

NDUFA12
Genomic Map