Assay Details
| Gene Symbol: SOCS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005585130.2 | XP_005585187.1 | 1 - 2 | 321 | 111 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005585130.2 transcripts.
Set Membership: |
|

SOCS3
Genomic Map