Assay Details
| Gene Symbol: NDUFA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283955.1 | NP_001270884.1 | 2 - 3 | 91 | 79 | |
| GenBank mRNA | AB169552.1 | - | 2 - 3 | 91 | 79 | |
More Information
Set Membership: |
|

NDUFA5
Genomic Map