Assay Details
| Gene Symbol: PPP1R7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284995.1 | NP_001271924.1 | 8 - 9 | 1030 | 67 | |
| XM_005574864.2 | XP_005574921.1 | 6 - 7 | 1427 | 67 | ||
| XM_015432095.1 | XP_015287581.1 | 7 - 8 | 1556 | 67 | ||
| XM_015432096.1 | XP_015287582.1 | 7 - 8 | 916 | 67 | ||
| GenBank mRNA | AB172271.1 | - | 8 - 9 | 1030 | 67 | |
More Information
Set Membership: |
|

PPP1R7
Genomic Map