Assay Details
| Gene Symbol: Chuk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162410.1 | NP_001155882.1 | 14 - 15 | 1656 | 103 | |
| NM_007700.2 | NP_031726.2 | 14 - 15 | 1656 | 103 | ||
| GenBank mRNA | AK018671.1 | - | 14 - 15 | 1635 | 103 | |
| AK035326.1 | - | 14 - 15 | 1613 | 103 | ||
| AK150039.1 | - | 14 - 15 | 1635 | 103 | ||
| AK155328.1 | - | 14 - 15 | 1636 | 103 | ||
| AK155393.1 | - | 14 - 15 | 1657 | 103 | ||
| AK164236.1 | - | 14 - 15 | 1657 | 103 | ||
| AK193721.1 | - | 3 - 4 | 273 | 103 | ||
| BC018243.1 | - | 14 - 15 | 1648 | 4511275 | 103 | |
| BC125266.1 | - | 3 - 4 | 255 | 103 | ||
| U12473.1 | - | 14 - 15 | 1624 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Chuk
Genomic Map