Assay Details
| Gene Symbol: Chuk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162410.1 | NP_001155882.1 | 15 - 16 | 1763 | 73 | |
| NM_007700.2 | NP_031726.2 | 15 - 16 | 1763 | 73 | ||
| GenBank mRNA | AK018671.1 | - | 15 - 16 | 1742 | 73 | |
| AK035326.1 | - | 15 - 16 | 1720 | 73 | ||
| AK150039.1 | - | 15 - 16 | 1742 | 73 | ||
| AK155328.1 | - | 15 - 16 | 1743 | 73 | ||
| AK155393.1 | - | 15 - 16 | 1764 | 73 | ||
| AK164236.1 | - | 15 - 16 | 1764 | 73 | ||
| BC018243.1 | - | 15 - 16 | 1755 | 4511275 | 73 | |
| BC125266.1 | - | 4 - 5 | 362 | 73 | ||
| U12473.1 | - | 15 - 16 | 1731 | 73 | ||
More Information
Set Membership: |
|

Chuk
Genomic Map