Assay Details
| Gene Symbol: Chuk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162410.1 | NP_001155882.1 | 16 - 17 | 1813 | 73 | |
| NM_007700.2 | NP_031726.2 | 16 - 17 | 1813 | 73 | ||
| GenBank mRNA | AK035326.1 | - | 16 - 17 | 1770 | 73 | |
| AK150039.1 | - | 16 - 17 | 1792 | 73 | ||
| AK155328.1 | - | 16 - 17 | 1793 | 73 | ||
| AK155393.1 | - | 16 - 17 | 1814 | 73 | ||
| AK164236.1 | - | 16 - 17 | 1814 | 73 | ||
| AK206454.1 | - | 2 - 3 | 75 | 73 | ||
| BC125266.1 | - | 5 - 6 | 412 | 73 | ||
| U12473.1 | - | 16 - 17 | 1781 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Chuk
Genomic Map